ENA159843578 Enamine

2-chloro-N-{[1-(dimethylamino)cyclohexyl]methyl}acetamide hydrochloride

  • MDL number MFCD09971353
  • Cas Number : 1172016-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30389
Availability details for
Quantity
  • Availability
  • Add to Cart