ENA159843581 Enamine

3-[(3-aminophenyl)methyl]imidazolidine-2,4-dione

  • MDL number MFCD09813883
  • Cas Number : 515143-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31884
Availability details for
Quantity
  • Availability
  • Add to Cart