ENA159843585 Enamine

4-methyl-6-oxo-2-sulfanyl-1,6-dihydropyridine-3-carbonitrile; morpholine

  • MDL number MFCD00716333
  • Cas Number : 93300-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data morpholine
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31893
Availability details for
Quantity
  • Availability
  • Add to Cart