ENA159843586 Enamine

(2-morpholin-4-ylpyridin-4-yl)methylamine

  • MDL number MFCD08060472
  • Cas Number : 864068-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31894

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31894
Availability details for
Quantity
  • Availability
  • Add to Cart