ENA159843587 Enamine

3-chloro-2-(1H-imidazol-1-yl)aniline

  • MDL number MFCD04971008
  • Cas Number : 869942-74-5
  • Molecular Weight : 193.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31895
Availability details for
Quantity
  • Availability
  • Add to Cart