ENA159843591 Enamine

2-(piperidin-4-yl)-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD02178927
  • Cas Number : 824403-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30444
Availability details for
Quantity
  • Availability
  • Add to Cart