ENA159843595 Enamine

2,5-dimethyl-1-(pyridin-2-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09733768
  • Cas Number : 953894-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31903
Availability details for
Quantity
  • Availability
  • Add to Cart