ENA159843601 Enamine

1-(3-methylbutanoyl)-1,4-diazepane

  • MDL number MFCD08444715
  • Cas Number : 923249-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31916

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31916
Availability details for
Quantity
  • Availability
  • Add to Cart