ENA159843604 Enamine

3-(4-ethyl-2,3-dioxopiperazin-1-yl)propanenitrile

  • MDL number MFCD09942989
  • Cas Number : 1017021-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31921

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31921
Availability details for
Quantity
  • Availability
  • Add to Cart