ENA159843610 Enamine

1-[(oxolan-2-yl)methyl]-1,4-diazepane

  • MDL number MFCD09040665
  • Cas Number : 926200-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31928
Availability details for
Quantity
  • Availability
  • Add to Cart