ENA159843612 Enamine

4-chloro-2-(1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD09732958
  • Cas Number : 55540-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31931
Availability details for
Quantity
  • Availability
  • Add to Cart