ENA159843616 Enamine

2-ethylbutanethioamide

  • MDL number MFCD09048499
  • Cas Number : 98278-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31936

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31936
Availability details for
Quantity
  • Availability
  • Add to Cart