ENA159843624 Enamine

1-(adamantan-1-yl)propan-1-amine

  • Cas Number : 60196-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31955
Availability details for
Quantity
  • Availability
  • Add to Cart