ENA159843625 Enamine

4-methylpentan-1-amine

  • MDL number MFCD06212593
  • Cas Number : 5344-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31956

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-31956
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart