ENA159843629 Enamine

4-(chloromethyl)-2-(2,4-dimethoxyphenyl)-1,3-thiazole

  • MDL number MFCD06655332
  • Cas Number : 900640-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30551
Availability details for
Quantity
  • Availability
  • Add to Cart