ENA159843637 Enamine

2-(1,2,3,4-tetrahydroisoquinolin-2-yl)aniline

  • MDL number MFCD07365122
  • Cas Number : 246247-91-6
  • Molecular Weight : 224.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31976
Availability details for
Quantity
  • Availability
  • Add to Cart