ENA159843640 Enamine

ethyl 6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD00176475
  • Cas Number : 17994-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31990
Availability details for
Quantity
  • Availability
  • Add to Cart