ENA159843642 Enamine

1-amino-3-(dimethylamino)propan-2-ol

  • MDL number MFCD09740503
  • Cas Number : 50411-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31994
Availability details for
Quantity
  • Availability
  • Add to Cart