ENA159843643 Enamine

N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD01712022
  • Cas Number : 64037-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30607
Availability details for
Quantity
  • Availability
  • Add to Cart