ENA159843644 Enamine

N-cyclohexyl-N-methylpiperazine-1-sulfonamide

  • MDL number MFCD08444721
  • Cas Number : 923156-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31996
Availability details for
Quantity
  • Availability
  • Add to Cart