ENA159843647 Enamine

2-(2-chloroacetamido)-N-(propan-2-yl)benzamide

  • MDL number MFCD03257530
  • Cas Number : 461649-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-32000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32000
Availability details for
Quantity
  • Availability
  • Add to Cart