ENA159843651 Enamine

2-(cyclopropylformamido)-4-(methylsulfanyl)butanoic acid

  • MDL number MFCD09045469
  • Cas Number : 65054-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32010
Availability details for
Quantity
  • Availability
  • Add to Cart