ENA159843657 Enamine

N-[3-(aminomethyl)phenyl]-2-methoxyacetamide

  • MDL number MFCD09044608
  • Cas Number : 926268-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32017
Availability details for
Quantity
  • Availability
  • Add to Cart