ENA159843664 Enamine

2,6-dichloro-3-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD08443169
  • Cas Number : 64063-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30729
Availability details for
Quantity
  • Availability
  • Add to Cart