ENA159843667 Enamine

2,6-dimethyl-4-(piperazine-1-sulfonyl)morpholine

  • MDL number MFCD09971609
  • Cas Number : 1042788-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32034
Availability details for
Quantity
  • Availability
  • Add to Cart