ENA159843670 Enamine

6,7-difluoroquinoline

  • MDL number MFCD01939054
  • Cas Number : 127827-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32038
Availability details for
Quantity
  • Availability
  • Add to Cart