ENA159843673 Enamine

3-(2-methyl-4-oxoquinazolin-3(4H)-yl)propanoic acid

  • MDL number MFCD07475561
  • Cas Number : 65452-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32041

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32041
Availability details for
Quantity
  • Availability
  • Add to Cart