ENA159843684 Enamine

4-phenylcyclohexanamine

  • MDL number MFCD00130038
  • Cas Number : 19992-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33153

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-33153
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart