ENA159843685 Enamine

N-[1-(2-methoxyphenyl)ethyl]-N-methylamine

  • MDL number MFCD01123285
  • Cas Number : 921074-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33215

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33215
Availability details for
Quantity
  • Availability
  • Add to Cart