ENA159843698 Enamine

(2-methoxyethyl)(2-methylpropyl)amine

  • MDL number MFCD09971623
  • Cas Number : 1019603-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33330
Availability details for
Quantity
  • Availability
  • Add to Cart