ENA159843707 Enamine

1-(1H-1,3-benzodiazol-2-yl)-3-(methylsulfanyl)propan-1-amine dihydrochloride

  • MDL number MFCD07209706
  • Cas Number : 1171352-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31229
Availability details for
Quantity
  • Availability
  • Add to Cart