ENA159843719 Enamine

2-(4-methyl-3-nitrophenyl)-1,3,4-oxadiazole

  • MDL number MFCD09971450
  • Cas Number : 460722-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31291
Availability details for
Quantity
  • Availability
  • Add to Cart