ENA159843724 Enamine

1-[4-(aminomethyl)phenyl]pyrrolidin-2-one hydrochloride

  • MDL number MFCD06799747
  • Cas Number : 1170201-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 267...269
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31343

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-31343
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart