ENA159843728 Enamine

6-[(2-aminoethyl)amino]pyridine-3-carbonitrile

  • MDL number MFCD09928078
  • Cas Number : 202460-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33381
Availability details for
Quantity
  • Availability
  • Add to Cart