ENA159843730 Enamine

2-(ethylamino)-N-(3-methoxyphenyl)acetamide

  • MDL number MFCD05263159
  • Cas Number : 743444-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31354
Availability details for
Quantity
  • Availability
  • Add to Cart