ENA159843733 Enamine

methyl 2-(4-acetamidophenoxy)acetate

  • MDL number MFCD03899149
  • Cas Number : 220299-99-0
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33396
Availability details for
Quantity
  • Availability
  • Add to Cart