ENA159843734 Enamine

N-[3-(3-cyclopropyl-5-sulfanyl-4H-1,2,4-triazol-4-yl)phenyl]acetamide

  • MDL number MFCD09971477
  • Cas Number : 1031130-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31393
Availability details for
Quantity
  • Availability
  • Add to Cart