ENA159843740 Enamine

2-(4-ethoxyphenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07375282
  • Cas Number : 147046-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31394
Availability details for
Quantity
  • Availability
  • Add to Cart