ENA159843755 Enamine

7-(4-chlorophenyl)-4,7-dioxoheptanoic acid

  • MDL number MFCD03488581
  • Cas Number : 1566-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33434
Availability details for
Quantity
  • Availability
  • Add to Cart