ENA159843759 Enamine

3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylic acid

  • MDL number MFCD00051658
  • Cas Number : 3919-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31438

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-31438
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart