ENA159843763 Enamine

1-(3-chloro-4-methylphenyl)-5-cyclopropyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971652
  • Cas Number : 1135055-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33444
Availability details for
Quantity
  • Availability
  • Add to Cart