ENA159843765 Enamine

2-chloroquinoline-4-carboxamide

  • MDL number MFCD01926028
  • Cas Number : 4295-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33447
Availability details for
Quantity
  • Availability
  • Add to Cart