ENA159843776 Enamine

[(cyanomethyl)thio]acetic acid

  • MDL number MFCD08705779
  • Cas Number : 55817-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33483
Availability details for
Quantity
  • Availability
  • Add to Cart