ENA159843780 Enamine

(butyrylamino)acetic acid

  • MDL number MFCD09812238
  • Cas Number : 20208-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31505

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31505
Availability details for
Quantity
  • Availability
  • Add to Cart