ENA159843783 Enamine

2-(thiophen-2-ylsulfanyl)acetic acid

  • MDL number MFCD01859892
  • Cas Number : 7342-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-33493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33493
Availability details for
Quantity
  • Availability
  • Add to Cart