ENA159843788 Enamine

3-(3-aminophenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD09971661
  • Cas Number : 34232-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33509

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33509
Availability details for
Quantity
  • Availability
  • Add to Cart