ENA159843792 Enamine

3-[(2-aminoethyl)amino]-1lambda6-thiolane-1,1-dione dihydrochloride

  • MDL number MFCD06799735
  • Cas Number : 329325-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33514
Availability details for
Quantity
  • Availability
  • Add to Cart