ENA159843796 Enamine

1-(4-fluorophenyl)-5-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971514
  • Cas Number : 1152975-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31546
Availability details for
Quantity
  • Availability
  • Add to Cart