ENA159843812 Enamine

2-{[(4-methoxy-3-nitrophenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD09971670
  • Cas Number : 22216-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-132
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-33552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33552
Availability details for
Quantity
  • Availability
  • Add to Cart