ENA159843816 Enamine

2-(2-chlorophenyl)-2-morpholin-4-ylethanamine

  • MDL number MFCD01631948
  • Cas Number : 866782-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33561

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33561
Availability details for
Quantity
  • Availability
  • Add to Cart